Order Entry
Puerto Rico
ContactUsLinkComponent
Cesium Ionization Buffer, 1%, for ICP, Inorganic Ventures
Cesium Ionization Buffer, 1%, for ICP, Inorganic Ventures
Catalog # 100359-758
Supplier:  Inorganic Ventures
CAS Number:  
undefined
Cesium Ionization Buffer, 1%, for ICP, Inorganic Ventures
Catalog # 100359-758
Supplier:  Inorganic Ventures
Supplier Number:  CSN-ISB-500ML
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Concentration:
    10000 µg/ml
  • Matrix:
    0.1% HNO₃
  • Pack type:
    Plastic Bottle
  • Size:
    500 ml
  • Cat. No.:
    100359-758
  • Element:
    Cesium
  • Supplier no.:
    CSNISB5

Specifications

About this item

Cesium ionization buffer, 1% for ICP is a reference material set in a nitric acid matrix for stability.

Certifications: Traceable to NIST. Acoording to ISO 17025, ISO 17034 and ISO 9001.