

bisbenzimide H-33342 trihydrochloride (Hoechst 33342) 10 mg/mL in water
Catalog # 89139-124
Supplier: Biotium
CAS Number:
Specifications
- Size:10 ml
- Cat. No.:89139-124
- Supplier no.:40046
Specifications
About this item
:
:561.93 g/mol
EINECS:245-691-1
:GHS07
:GHS08
Storage Temperature:Refrigerator
CAS Number:23491-52-3
:MFCD00012679
Formula:C₂₇H₂₈N₆O·3HCl