bisBenzimide H 33258 trihydrochloride (Hoechst 33258) 10 mg/mL in water
Catalog # 89139-120
Supplier: Biotium
CAS Number:
Specifications
- Size:10 ml
- Cat. No.:89139-120
- Supplier no.:40044
Specifications
About this item
:
:533.88 g/mol
Storage Temperature:Refrigerator
EINECS:245-690-6
CAS Number:23491-45-4
Formula:C₂₅H₂₄N₆O·3HCl
MDL Number:MFCD00012679