Sulfuric acid 95.0-97.0%, Puriss. p.a. ACS, ISO, Reag. Ph. Eur. for the determination of mercury, Fluka™
:7664-93-9
:
:H₂SO₄
:MFCD00064589
:98.08 g/mol
:310 °C (1.013 hPa)
:–14…–10 °C
:1.840 g/cm³ (20 °C)
:Ambient
:231-639-5
:1830
:8,II
:GHS05
- Vapor Pressure:1 mmHg ( 146 °C)
- Assay:95.0 - 97.0 %
- Residue on ignition:Max. 5 ppm
- Ammonium (NH₄):Max. 2 ppm
- Silver (Ag):Max. 0.000002 %
- Aluminium (Al):Max. 0.000005 %
- Arsenic (As):Max. 0.01 ppm
- Barium (Ba):Max. 0.000005 %
- Beryllium (Be):Max. 0.000001 %
- Bismuth (Bi):Max. 0.000005 %
- Calcium (Ca):Max. 0.00002 %
- Cadmium (Cd):Max. 0.000002 %
- Cobalt (Co):Max. 0.000001 %
- Chromium (Cr):Max. 0.000005 %
- Copper (Cu):Max. 0.000001 %
- Iron (Fe):Max. 0.1 ppm
- Germanium (Ge):Max. 0.000005 %
- Mercury (Hg):Max. 0.005 ppm
- Potassium (K):Max. 0.00001 %
- Lithium (Li):Max. 0.000001 %
- Magnesium (Mg):Max. 0.000005 %
- Manganese (Mn):Max. 0.000001 %
- Molybdenum (Mo):Max. 0.000002 %
- Sodium (Na):Max. 0.00005 %
- Nickel (Ni):Max. 0.000002 %
- Lead (Pb):Max. 0.000002 %
- Strontium (Sr):Max. 0.000002 %
- Titanium (Ti):Max. 0.00001 %
- Thallium (Tl):Max. 0.000005 %
- Vanadium (V):Max. 0.000001 %
- Zinc (Zn):Max. 0.000005 %
- Zirconium (Zr):Max. 0.00001 %
- Chloride (Cl):Max. 0.1 ppm
- Nitrate (NO₃):Max. 0.2 ppm
- Phosphate (PO₄):Max. 0.00005 %
- KMnO₄ reducing matter (as O):Max. 0.0002 %
- APHA:Max. 10
- KMnO₄ reducing matter (as SO₂):Max. 2 ppm