Order Entry
Puerto Rico
ContactUsLinkComponent
Sodium thiosulfate pentahydrate 99-101%, Puriss., meets analytical specification of BP, Ph. Eur., USP, Fluka™
Sodium thiosulfate pentahydrate 99-101%, Puriss., meets analytical specification of BP, Ph. Eur., USP, Fluka™
Catalog # BJ13479-500G
CAS Number:  
Sodium thiosulfate pentahydrate 99-101%, Puriss., meets analytical specification of BP, Ph. Eur., USP, Fluka™
Catalog # BJ13479-500G
Supplier Number:  13479-500G
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    500 g
  • Cat. No.:
    BJ13479-500G
  • Supplier no.:
    13479-500G

Specifications

Specification Test Results

  • Identity:
    complying
  • Assay:
    99.0 - 101.0 %
  • Assay (calculated on the anhydrous basis):
    99.0 - 100.5 %
  • Loss on drying (vacuum, 16 h):
    32.0 - 37.0 %
  • pH (10 %, 20°C):
    6.0 - 9.0
  • Arsenic (As):
    Max. 0.0003 %
  • Calcium (Ca):
    Max. 0.005 %
  • Iron (Fe):
    Max. 0.001 %
  • Heavy metals (as Pb):
    Max. 0.0005 %
  • Chloride (Cl):
    Max. 0.05 %
  • Sulfate and sulfite (as SO₄):
    Max. 0.2 %
  • Sulfide (S):
    complying
  • Appearance of the solution:
    complying
  • Residual solvents:
    complying

Specification Test Results

About this item

 :
 :248.19 g/mol
Boiling Pt: Dec. (on heating)
Melting Pt:48 °C (dec.)
Density:1.73 g/cm³ (20 °C)
Storage Temperature:Room Temperature
CAS Number:10102-17-7
Formula:Na₂S₂O₃.5H₂O
MDL Number:MFCD00149186