Sodium thiosulfate 1 M Na2S2O3 (1N) volumetric solution, Fluka™
Catalog # BJ35244-1L
Supplier: Honeywell Research Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Size:1 L
- Cat. No.:BJ35244-1L
Specifications
Specification Test Results
- Factor:0.999 - 1.001
Specification Test Results
About this item
:
:158.11 g/mol
Boiling Pt:100 °C (1.013 hPa)
Density:1.120 g/cm³ (20 °C)
Storage Temperature:Ambient
CAS Number:7772-98-7
Formula:Na₂S₂O₃
MDL Number:MFCD00003499