Sodium thiosulfate 0.2 M Na2S2O3 (0.2N) volumetric solution, Fluka™
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:7772-98-7
:
Formula:Na₂S₂O₃
MDL Number:MFCD00003499
:158.11 g/mol
Boiling Pt:100 °C
Density:1.01 g/cm³
Storage Temperature:Ambient
Specification Test Results
- Factor:0.998 - 1.002