Order Entry
Puerto Rico
ContactUsLinkComponent
Sodium molybdate(VI) dihydrate ≥99% ACS, Fluka™
Sodium molybdate(VI) dihydrate ≥99% ACS, Fluka™
Certificates
Sodium molybdate(VI) dihydrate ≥99% ACS, Fluka™
Certificates

Specification Test Results

  • Assay:
    99.5 - 103.0 %
  • pH (5 %, 25 °C):
    7.0 - 10.5
  • Water insoluble matter:
    Max. 0.005 %
  • Iron (Fe):
    Max. 0.001 %
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Chloride (Cl):
    Max. 0.005 %
  • Ammonium (NH₄):
    Max. 0.001 %
  • Phosphate (PO₄):
    Max. 5 ppm
  • Sulfate (SO₄):
    Max. 0.015 %

Specification Test Results

Related Information