
Sodium hydroxide, anhydrous ≥98%, pellets, Reagent Grade
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
:40 g/mol
Boiling Pt:1.390 °C (1.013 hPa)
Melting Pt:319 °C
Density:2.13 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1823
ADR:8,II
:GHS05
Specification Test Results
- Vapor Density:>1 (vs air)
- Assay:Min. 98.0 %
- Assay of Na₂CO₃:Max. 1.0 %
- Appearance of the solution:complying
- Turbidity of solution:Max. 3.5 NTU
- Vapor Pressure:3 mmHg ( 37 °C)