Order Entry
Puerto Rico
ContactUsLinkComponent
Sodium formate solution 10 mM NaOH in 2-propanol/water (1:1) (+0.2% HCOOH) for LC-MS, Fluka™
Flammable
Irritant
Sodium formate solution 10 mM NaOH in 2-propanol/water (1:1) (+0.2% HCOOH) for LC-MS, Fluka™
Catalog # BJ97574-100ML
CAS Number:  
Flammable
Irritant
Sodium formate solution 10 mM NaOH in 2-propanol/water (1:1) (+0.2% HCOOH) for LC-MS, Fluka™
Catalog # BJ97574-100ML
Supplier Number:  97574-100ML
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    100 ml
  • Cat. No.:
    BJ97574-100ML

Specifications

Specification Test Results

  • Identity (LC-MS):
    complying
  • LC-MS application:
    complying
  • Suitability for the LC-MS:
    complying

Specification Test Results

About this item

 :
 :68.01 g/mol
Flash Pt:22 °C
Storage Temperature:Ambient
UN:1219
ADR:3,II
 :GHS02
 :GHS07
CAS Number:141-53-7
Formula:HCO₂Na
MDL Number:MFCD00013101