Order Entry
Puerto Rico
ContactUsLinkComponent
Sodium carbonate concentrate for dilution to 1L, 0.05 M Na2CO3 (0.1N) volumetric solution, Fluka™
Irritant
Sodium carbonate concentrate for dilution to 1L, 0.05 M Na2CO3 (0.1N) volumetric solution, Fluka™
Catalog # BJ38170-1EA
CAS Number:  
undefined
Irritant
Sodium carbonate concentrate for dilution to 1L, 0.05 M Na2CO3 (0.1N) volumetric solution, Fluka™
Catalog # BJ38170-1EA
Supplier Number:  38170-1EA
CAS Number:  

Specifications

  • Pack type:
    Ampoule
  • Size:
    1 Each
  • Cat. No.:
    BJ38170-1EA

Specifications

Specification Test Results

  • Factor:
    0.998 - 1.002
  • BAM/EMPA reference material:
    effective
  • BAM/EMPA Certificate Number:
    effective
  • Traceable to NIST SRM:
    effective

Specification Test Results

About this item

 :
 :105.99 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:497-19-8
Formula:Na₂CO₃
MDL Number:MFCD00003494