
Potassium nitrate ≥99%, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Fluka™
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:7757-79-1
:
Formula:KNO₃
MDL Number:MFCD00011409
:101.1 g/mol
Boiling Pt: Dec.
Melting Pt:335 °C (1.013 hPa)
Density:2.11 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-818-8
UN:1486
ADR:5.1,III
:GHS03
Specification Test Results
- Assay:Min. 99.0 %
- pH (5 %, 25 °C):5.0 - 8.0
- Water insoluble matter:Max. 0.005 %
- Ammonium (NH₄):Max. 0.001 %
- Calcium (Ca):Max. 0.001 %
- Copper (Cu):Max. 0.0005 %
- Iron (Fe):Max. 2 ppm
- Magnesium (Mg):Max. 0.002 %
- Sodium (Na):Max. 0.005 %
- Lead (Pb):Max. 0.0005 %
- Zinc (Zn):Max. 0.0005 %
- Heavy metals (as Pb):Max. 5 ppm
- Chloride (Cl):Max. 0.001 %
- Iodate (IO₃):Max. 5 ppm
- Nitrite (NO₂):Max. 0.001 %
- Phosphate (PO₄):Max. 5 ppm
- Sulfate (SO₄):Max. 0.002 %