Potassium hexacyanoferrate(III) 0.1 M K3Fe(CN)6 in aqueous solution volumetric solution, Fluka™
BJ34272-1L
: Avantor
:
- Pack type:Plastic bottle
- Size:1 L
- Cat. No.:BJ34272-1L
- Factor:0.99 - 1.01
:
:329.25 g/mol
:1 g/cm3 (20 °C)
:Ambient
:13746-66-2
:K₃Fe(CN)₆
:MFCD00011392