Order Entry
Puerto Rico
ContactUsLinkComponent
Potassium fluoride ≥99.0% (fusing), Purum p.a.
Corrosive
Toxic
Potassium fluoride ≥99.0% (fusing), Purum p.a.
Certificates
Corrosive
Toxic
Potassium fluoride ≥99.0% (fusing), Purum p.a.

Specification Test Results

  • Assay:
    99.0 - 101.0 %
  • Calcium (Ca):
    Max. 50 ppm
  • Cadmium (Cd):
    Max. 50 ppm
  • Cobalt (Co):
    Max. 50 ppm
  • Copper (Cu):
    Max. 50 ppm
  • Iron (Fe):
    Max. 50 ppm
  • Sodium (Na):
    Max. 5000 ppm
  • Nickel (Ni):
    Max. 50 ppm
  • Lead (Pb):
    Max. 50 ppm
  • Zinc (Zn):
    Max. 50 ppm
  • Total S (as SO₄):
    Max. 1000 ppm
  • Chloride (Cl):
    Max. 50 ppm

Specification Test Results

Related Information