Order Entry
Puerto Rico
ContactUsLinkComponent
Potassium dihydrogen phosphate, anhydrous 98-100.5% (dried basis), meets analytical specification of NF, Ph. Eur., E340, Fluka™
Potassium dihydrogen phosphate, anhydrous 98-100.5% (dried basis), meets analytical specification of NF, Ph. Eur., E340, Fluka™
Certificates
Potassium dihydrogen phosphate, anhydrous 98-100.5% (dried basis), meets analytical specification of NF, Ph. Eur., E340, Fluka™
Certificates

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specification Test Results

  • Assay (calculated to the dried substance):
    98.0 - 100.5 %
  • Loss on drying (130°C):
    Max. 1.0 %
  • Loss on drying (105°C, 4 h):
    Max. 1.0 %
  • Water insoluble matter:
    Max. 0.2 %
  • pH (5 %, 20°C):
    4.2 - 4.5
  • Arsenic (As):
    Max. 1 ppm
  • Cadmium (Cd):
    Max. 1 ppm
  • Copper (Cu):
    Max. 0.002 %
  • Iron (Fe):
    Max. 10 ppm
  • Mercury (Hg):
    Max. 1 ppm
  • Sodium (Na):
    Max. 0.1 %
  • Lead (Pb):
    Max. 1 ppm
  • Zinc (Zn):
    Max. 0.002 %
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Chloride (Cl):
    Max. 10 ppm
  • Fluoride (F):
    Max. 10 ppm
  • Sulfate (SO₄):
    Max. 200 ppm
  • KMnO₄ reducing matter (as O):
    Max. 0.04 %
  • Appearance of the solution:
    complying
  • Residual solvents:
    complying

Specification Test Results