Order Entry
Puerto Rico
ContactUsLinkComponent
Potassium bromate 1/60 M KBrO3 (0.1N) volumetric solution, Fluka™
Health
Potassium bromate 1/60 M KBrO3 (0.1N) volumetric solution, Fluka™
Catalog # BJ34268-1L
CAS Number:  
Health
Potassium bromate 1/60 M KBrO3 (0.1N) volumetric solution, Fluka™
Catalog # BJ34268-1L
Supplier Number:  34268-1L
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    1 L
  • Cat. No.:
    BJ34268-1L

Specifications

Specification Test Results

  • Assay:
    0.01663 - 0.01670 mol/l

Specification Test Results

About this item

 :
 :167 g/mol
 :GHS08
Storage Temperature:Ambient
CAS Number:7758-01-2
Formula:KBrO₃
MDL Number:MFCD00011359