Order Entry
Puerto Rico
ContactUsLinkComponent
Orthophosphoric acid in water ≥85% (w/w) ACS, Fluka™
Corrosive
Orthophosphoric acid in water ≥85% (w/w) ACS, Fluka™
Certificates
Corrosive
Orthophosphoric acid in water ≥85% (w/w) ACS, Fluka™
Certificates

Specification Test Results

  • Assay:
    Min. 85.0 %
  • Volatile acids (CH₃COOH):
    Max. 0.001 %
  • Water insoluble matter:
    Max. 0.001 %
  • Arsenic (As):
    Max. 1 ppm
  • Calcium (Ca):
    Max. 0.002 %
  • Iron (Fe):
    Max. 0.003 %
  • Potassium (K):
    Max. 0.005 %
  • Magnesium (Mg):
    Max. 0.002 %
  • Manganese (Mn):
    Max. 0.5 ppm
  • Sodium (Na):
    Max. 0.025 %
  • Antimony (Sb):
    Max. 0.002 %
  • Heavy metals (as Pb):
    Max. 0.001 %
  • Chloride (Cl):
    Max. 3 ppm
  • Nitrate (NO₃):
    Max. 5 ppm
  • Sulfate (SO₄):
    Max. 0.003 %
  • Reducing impurities:
    complying
  • APHA:
    Max. 10
  • Vapor Density:
    3.4 (vs air)

Specification Test Results