Order Entry
Puerto Rico
ContactUsLinkComponent
 
Health
Irritant
N-Methyl-2-pyrrolidone (NMP) ≥99.5%, BioSyn™ for biosynthesis, for applications requiring low water content, Burdick & Jackson™
Catalog # BJBN305-20
Supplier:  Burdick & Jackson
CAS Number:  
undefined
Health
Irritant
N-Methyl-2-pyrrolidone (NMP) ≥99.5%, BioSyn™ for biosynthesis, for applications requiring low water content, Burdick & Jackson™
Catalog # BJBN305-20
Supplier:  Burdick & Jackson
Supplier Number:  BN305-20
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    NOWPAK® II, returnable
  • Size:
    20 L
  • Cat. No.:
    BJBN305-20
  • Supplier no.:
    BN305-20

Specifications

Specification Test Results

  • Amines (as dimethylamine):
    3 ppm max.
  • GC Analysis:
    99.5 % min.
  • Refractive Index (20ºC):
    1.4680 - 1.4720
  • Residue:
    5 mg/L max.
  • UV Absorbance @ 300 nm:
    0.5 AU max.
  • UV Absorbance @ 325 nm:
    0.1 AU max.
  • UV Cutoff:
    285 nm max.
  • Water by Karl Fischer Titration:
    200 ppm max.

Specification Test Results

About this item

 :
 :99.13 g/mol
Boiling Pt:202 °C
Melting Pt:–24 °C
Density:1.0259 g/cm3
Flash Pt:88 °C
 :GHS08
 :GHS07
Storage Temperature:Ambient
CAS Number:872-50-4
Formula:C₅H₉NO
MDL Number:MFCD00003193