Order Entry
Puerto Rico
ContactUsLinkComponent
 
 
 
N-Methyl-2-pyrrolidone (NMP) ≥99.0% ACS, Riedel-de Haën™
 
undefined
 
 
N-Methyl-2-pyrrolidone (NMP) ≥99.0% ACS, Riedel-de Haën™
 

 

  • Assay (GC):
    Min. 99.0 %
  • Water (Karl Fischer):
    Max. 0.05 %
  • Free amine:
    Max. 0.01 %
  • Chloride (Cl):
    Max. 0.0001 %
  • APHA:
    Max. 50
  • IR spectrum:
    complying
  • Vapor Density:
    3.4 (vs air)
  • Vapor Pressure:
    0.99 mmHg ( 40 °C)
  • Autoignition Temperature:
    518 °F