Molecular sieve 3A (0.3 nm, 3 Å) ≥99.5%, rods 1/16 in., Fluka™
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:308080-99-1
:1318-02-1
Formula:KnNa₁₂-n[(AlO₂)₁₂(SiO₂)₁₂]·xH₂O
MDL Number:MFCD00131613
Melting Pt:400 °C
Density:2 g/cm³
Storage Temperature:Ambient
Specification Test Results
- Size:1/16 in.
- Granulation:complying
- Pore Size (Ång Ström):3
- Loss on ignition (900°C):Max. 3 %