Molecular sieve 3A (0.3 nm, 3 Å), pellets, Diameter 3.2 mm, Fluka™
BJ334278-1KG
:
- Pack type:Plastic bottle
- Size:1 kg
- Cat. No.:BJ334278-1KG
- Particle Size:3.2 mm
- Form:pellets
- Granulation:complying
- Pore Size (Ång Ström):3
:
:400 °C
:2 g/cm³
:Ambient
:308080-99-1
:1318-02-1
:KnNa₁₂-n[(AlO₂)₁₂(SiO₂)₁₂]·xH₂O
:MFCD00131613