Order Entry
Puerto Rico
ContactUsLinkComponent
Mercury(II) nitrate 0.005 M Hg(NO3)2 (0.01N) ≥98% volumetric solution, for chloride determination, Fluka™
Health
Mercury(II) nitrate 0.005 M Hg(NO3)2 (0.01N) ≥98% volumetric solution, for chloride determination, Fluka™
Catalog # BJ34289-1L
CAS Number:  
Health
Mercury(II) nitrate 0.005 M Hg(NO3)2 (0.01N) ≥98% volumetric solution, for chloride determination, Fluka™
Catalog # BJ34289-1L
Supplier Number:  34289-1L
CAS Number:  

Specifications

  • Pack type:
    Glass Bottle
  • Size:
    1 L
  • Cat. No.:
    BJ34289-1L

Specifications

Specification Test Results

  • Factor:
    0.995 - 1.005

Specification Test Results

About this item

 :
 :324.6
Boiling Pt:100 °C (solvent)
Melting Pt:<0 °C
Density:1.0 g/cm³ (20 °C)
Flash Pt:Does not flash
 :GHS08
Storage Temperature:Ambient
CAS Number:10045-94-0
Formula:HgN₂O₆
MDL Number:MFCD00011038
Flash Pt: Does not flash