Magnesium sulfate, anhydrous ≥98.0% (by complexometric analysis), extra fine powder, Puriss. p.a. drying agent, Fluka™
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:7487-88-9
:
Formula:MgSO₄
MDL Number:MFCD00011110
:120.37 g/mol
Melting Pt:1.130 °C (dec.)
Density:2.7 g/cm³
Storage Temperature:Ambient
EINECS:231-298-2
Specification Test Results
- Form:powder (very fine)
- Assay:98.0 - 102.0 %
- Loss on ignition (600°C):Max. 2 %
- Aluminium (Al):Max. 20 ppm
- Arsenic (As):Max. 0.1 ppm
- Barium (Ba):Max. 5 ppm
- Bismuth (Bi):Max. 5 ppm
- Calcium (Ca):Max. 400 ppm
- Cadmium (Cd):Max. 5 ppm
- Cobalt (Co):Max. 5 ppm
- Chromium (Cr):Max. 5 ppm
- Copper (Cu):Max. 5 ppm
- Iron (Fe):Max. 10 ppm
- Potassium (K):Max. 200 ppm
- Lithium (Li):Max. 5 ppm
- Manganese (Mn):Max. 20 ppm
- Molybdenum (Mo):Max. 5 ppm
- Sodium (Na):Max. 100 ppm
- Nickel (Ni):Max. 5 ppm
- Lead (Pb):Max. 5 ppm
- Strontium (Sr):Max. 5 ppm
- Zinc (Zn):Max. 5 ppm
- Chloride (Cl):Max. 50 ppm
- Nitrate (NO₃):Max. 40 ppm
- Ammonium (NH₄):Max. 20 ppm