Magnesium acetate tetrahydrate ≥99%, Reagent Grade
Catalog # BJM0631-1KG
Supplier: Honeywell Research Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Size:1 kg
- Cat. No.:BJM0631-1KG
Specifications
Specification Test Results
- Assay:99 - 102 %
- Insoluble in acetic acid:Max. 0.005 %
- Form:Solid
Specification Test Results
About this item
:
:214.46 g/mol
Melting Pt:68 °C
Density:1.45 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:205-554-9
CAS Number:16674-78-5
Formula:(H₃CCOO)₂Mg·4H₂O
MDL Number:MFCD00149214