L(+)-Ascorbic acid ≥99.9998% (metals basis), TraceSELECT™, Fluka™
Catalog # BJ05878-100G
Supplier: Honeywell Research Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Size:100 g
- Cat. No.:BJ05878-100G
Specifications
Specification Test Results
- Ammonium (NH₄):Max. 5 ppm
- Assay:Min. 99.0 %
- Specific rotation (a) 20/D; c=10 in H₂O:20.5 - 21.5 °
- Silver (Ag):Max. 0.01 ppm
- Aluminium (Al):Max. 0.05 ppm
- Arsenic (As):Max. 0.005 ppm
- Gold (Au):Max. 0.01 ppm
- Barium (Ba):Max. 0.01 ppm
- Beryllium (Be):Max. 0.01 ppm
- Bismuth (Bi):Max. 0.01 ppm
- Calcium (Ca):Max. 0.1 ppm
- Cadmium (Cd):Max. 0.01 ppm
- Cerium (Ce):Max. 0.01 ppm
- Cobalt (Co):Max. 0.01 ppm
- Chromium (Cr):Max. 0.05 ppm
- Cesium (Cs):Max. 0.01 ppm
- Copper (Cu):Max. 0.01 ppm
- Dysprosium (Dy):Max. 0.01 ppm
- Erbium (Er):Max. 0.01 ppm
- Europium (Eu):Max. 0.01 ppm
- Iron (Fe):Max. 0.1 ppm
- Gallium (Ga):Max. 0.01 ppm
- Gadolinium (Gd):Max. 0.01 ppm
- Germanium (Ge):Max. 0.05 ppm
- Hafnium (Hf):Max. 0.01 ppm
- Mercury (Hg):Max. 0.01 ppm
- Holmium (Ho):Max. 0.01 ppm
- Indium (In):Max. 0.01 ppm
- Iridium (Ir):Max. 0.01 ppm
- Potassium (K):Max. 0.1 ppm
- Lanthanum (La):Max. 0.01 ppm
- Lithium (Li):Max. 0.01 ppm
- Lutetium (Lu):Max. 0.01 ppm
- Magnesium (Mg):Max. 0.05 ppm
- Manganese (Mn):Max. 0.01 ppm
- Molybdenum (Mo):Max. 0.01 ppm
- Sodium (Na):Max. 2 ppm
- Niobium (nb):Max. 0.01 ppm
- Neodymium (nd):Max. 0.01 ppm
- Nickel (Ni):Max. 0.01 ppm
- Lead (Pb):Max. 0.01 ppm
- Palladium (Pd):Max. 0.01 ppm
- Praseodymium (Pr):Max. 0.01 ppm
- Platinum (Pt):Max. 0.01 ppm
- Rubidium (Rb):Max. 0.01 ppm
- Rhenium (Re):Max. 0.01 ppm
- Rhodium (Rh):Max. 0.01 ppm
- Ruthenium (Ru):Max. 0.01 ppm
- Antimony (Sb):Max. 0.01 ppm
- Scandium (Sc):Max. 0.05 ppm
- Selenium (Se):Max. 0.01 ppm
- Samarium (Sm):Max. 0.01 ppm
- Tin (Sn):Max. 0.01 ppm
- Strontium (Sr):Max. 0.01 ppm
- Tantalum (Ta):Max. 0.01 ppm
- Terbium (Tb):Max. 0.01 ppm
- Tellurium (Te):Max. 0.01 ppm
- Thorium (Th):Max. 0.01 ppm
- Titanium (Ti):Max. 0.05 ppm
- Thallium (Tl):Max. 0.01 ppm
- Thulium (Tm):Max. 0.01 ppm
- Uran (U):Max. 0.01 ppm
- Vanadium (V):Max. 0.01 ppm
- Tungsten (W):Max. 0.01 ppm
- Yttrium (Y):Max. 0.01 ppm
- Ytterbium (Yb):Max. 0.01 ppm
- Zinc (Zn):Max. 0.05 ppm
- Zirconium (Zr):Max. 0.01 ppm
- Chloride (Cl):Max. 50 ppm
- Sulfate (SO₄):Max. 20 ppm
Specification Test Results
About this item
:
:176.13 g/mol
Melting Pt:190…194 °C
Density:1.65 g/cm³
Storage Temperature:Ambient
EINECS:200-066-2
CAS Number:50-81-7
Formula:C₆H₈O₆
MDL Number:MFCD00064328