Order Entry
Puerto Rico
ContactUsLinkComponent
 
 
 
Iron(III) chloride hexahydrate 97% ACS, Fluka™
 
undefined
 
 
Iron(III) chloride hexahydrate 97% ACS, Fluka™
 

 

  • Assay:
    97.0 - 102.0 %
  • Insoluble in acid:
    Max. 0.01 %
  • Calcium (Ca):
    Max. 0.01 %
  • Copper (Cu):
    Max. 0.003 %
  • Potassium (K):
    Max. 0.005 %
  • Magnesium (Mg):
    Max. 0.005 %
  • Sodium (Na):
    Max. 0.05 %
  • Zinc (Zn):
    Max. 0.003 %
  • Iron (Fe 2⁺):
    Max. 0.002 %
  • Nitrate (NO₃):
    Max. 0.01 %
  • Total phosphorus (as PO₄):
    Max. 0.01 %
  • Sulfate (SO₄):
    Max. 0.01 %

 

Frequently Bought Together