Order Entry
Puerto Rico
ContactUsLinkComponent
Hexane (mixture of isomers), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Hexane (mixture of isomers), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
  BJ34493-2.5L
 :  
undefined
Hexane (mixture of isomers), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
  BJ34493-2.5L
 :  34493-2.5L
 :  

 

  • Pack type:
    Glass bottle
  • Size:
    2.5 L
  • Cat. No.:
    BJ34493-2.5L

 

 

  • Assay of isomers (GC):
    Min. 85 %
  • Non-volatile matter:
    Max. 0.0005 %
  • Water (Karl Fischer):
    Max. 0.01 %
  • Suitability for residue analysis:
    Max. 5 ng/l
  • n-Hexane (GC):
    Max. 4.9 %
  • Explosion Limit:
    7.5 %
  • Autoignition Temperature:
    437 °F

 

 

 :
 :86.18 g/mol
 :48…65 °C (1.013 hPa)
 :–154 °C
 :0.66 g/cm³ (20 °C)
 :–26 °C
 :1208
 :3,II
 :GHS02
 :GHS08
 :GHS07
 :Ambient
 :110-54-3
 :92112-69-1
 :H₃C(CH₂)₄CH₃
 :MFCD02179311