Order Entry
Puerto Rico
ContactUsLinkComponent
 
di-Potassium hydrogen phosphate, anhydrous ≥99.0% (by titrimetric analysis), Puriss. p.a. ACS, Fluka™
Catalog # BJ60356-1KG
CAS Number:  
undefined
di-Potassium hydrogen phosphate, anhydrous ≥99.0% (by titrimetric analysis), Puriss. p.a. ACS, Fluka™
Catalog # BJ60356-1KG
Supplier Number:  60356-1KG
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    1 kg
  • Cat. No.:
    BJ60356-1KG
  • Supplier no.:
    60356-1KG

Specifications

Specification Test Results

  • Assay:
    99.0 - 101.0 %
  • Water insoluble matter:
    Max. 0.01 %
  • Loss on drying (105°C):
    Max. 1.0 %
  • pH (5 %, 25 °C):
    8.5 - 9.6
  • Calcium (Ca):
    Max. 50 ppm
  • Cadmium (Cd):
    Max. 5 ppm
  • Cobalt (Co):
    Max. 5 ppm
  • Chromium (Cr):
    Max. 5 ppm
  • Copper (Cu):
    Max. 5 ppm
  • Iron (Fe):
    Max. 10 ppm
  • Magnesium (Mg):
    Max. 5 ppm
  • Manganese (Mn):
    Max. 5 ppm
  • Sodium (Na):
    Max. 500 ppm
  • Nickel (Ni):
    Max. 5 ppm
  • Lead (Pb):
    Max. 5 ppm
  • Zinc (Zn):
    Max. 5 ppm
  • Chloride (Cl):
    Max. 30 ppm
  • Total S (as SO₄):
    Max. 50 ppm
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Total nitrogen (as N):
    Max. 10 ppm
  • Solution 5 g + 95 mL (H₂O):
    complying

Specification Test Results

About this item

 :
 :174.18 g/mol
Melting Pt:340 °C (dec.)
Storage Temperature:Ambient
EINECS:231-834-5
CAS Number:7758-11-4
Formula:K₂HPO₄
MDL Number:MFCD00011383