Order Entry
Puerto Rico
ContactUsLinkComponent
 
 
Copper(II) sulfate, anhydrous 99-100.5% (by anhydrous basis), Puriss., meets analytical specification of BP, Ph. Eur., USP, Fluka™
New Product
 
 
Copper(II) sulfate, anhydrous 99-100.5% (by anhydrous basis), Puriss., meets analytical specification of BP, Ph. Eur., USP, Fluka™
New Product
 

 

  • Assay (calculated to the dried substance):
    99.0 - 100.5 %
  • Loss on drying (250°C):
    Max. 1.0 %
  • Calcium (Ca):
    Max. 0.005 %
  • Iron (Fe):
    Max. 30 ppm
  • Potassium (K):
    Max. 0.01 %
  • Magnesium (Mg):
    Max. 0.01 %
  • Sodium (Na):
    Max. 0.02 %
  • Nickel (Ni):
    Max. 0.005 %
  • Lead (Pb):
    Max. 5 ppm
  • Chloride (Cl):
    Max. 100 ppm
  • Residual solvents:
    complying
  • Appearance of the solution:
    complying

 

Frequently Bought Together