

Cobalt(II) chloride hexahydrate 98-102%, Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™
Catalog # BJ31277-100G
Supplier: Honeywell Research Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Size:100 g
- Cat. No.:BJ31277-100G
- Supplier no.:31277-100G
Specifications
Specification Test Results
- Assay:98.0 - 102.0 %
- Water insoluble matter:Max. 0.01 %
- Calcium (Ca):Max. 0.005 %
- Iron (Fe):Max. 0.005 %
- Potassium (K):Max. 0.005 %
- Copper (Cu):Max. 0.002 %
- Magnesium (Mg):Max. 0.001 %
- Manganese (Mn):Max. 0.005 %
- Sodium (Na):Max. 0.01 %
- Nickel (Ni):Max. 0.05 %
- Lead (Pb):Max. 0.0005 %
- Zinc (Zn):Max. 0.005 %
- Nitrate (NO₃):Max. 0.01 %
- Sulfate (SO₄):Max. 0.005 %
Specification Test Results
About this item
:
:237.93 g/mol
Boiling Pt: Dec. (on heating)
Melting Pt:86 °C
Density:1.9 g/cm³ (20 °C)
:GHS07
:GHS08
Storage Temperature:Ambient
CAS Number:7791-13-1
Formula:CoCl₂·6H₂O
MDL Number:MFCD00149652