Calcium hydrogen orthophosphate, anhydrous ≥98.5% (by complexometric analysis), puriss. p.a., Fluka™
BJ21177-250G
:
- Pack type:Plastic bottle
- Size:250 g
- Cat. No.:BJ21177-250G
- Assay:98.5 - 101.0 %
- Loss on ignition (850°C):6.5 - 8.5 %
- Insoluble in hydrochloric acid:max. 0.2 %
- Arsenic (As):max. 0.5 ppm
- Barium (Ba):max. 10 ppm
- Cadmium (Cd):max. 5 ppm
- Cobalt (Co):max. 5 ppm
- Chromium (Cr):max. 5 ppm
- Copper (Cu):max. 5 ppm
- Iron (Fe):max. 200 ppm
- Potassium (K):max. 50 ppm
- Magnesium (Mg):max. 2000 ppm
- Manganese (Mn):max. 100 ppm
- Sodium (Na):max. 500 ppm
- Nickel (Ni):max. 5 ppm
- Lead (Pb):max. 5 ppm
- Strontium (Sr):max. 200 ppm
- Zinc (Zn):max. 5 ppm
- Total S (as SO₄):max. 1000 ppm
- Chloride (Cl):max. 100 ppm
:
:136.06 g/mol
:>450 °C
:Ambient
:231-826-1
:7757-93-9
:CaHPO₄
:MFCD00010909