Bismuth(III) nitrate basic 71-74% (Bi basis (calc. on dried substance)), Puriss., meets analytical specification of DAB, USP, Fluka™
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:1304-85-4
:
Formula:Bi₅O(OH)₉(NO₃)₄
MDL Number:MFCD00064844
:1461.99 g/mol
Melting Pt:>500 °C (dec.)
Density:5.29 g/cm³ (22.8 °C)
Storage Temperature:Ambient
EINECS:215-136-8
Specification Test Results
- Assay of Bi (calculated to dried substances):71 - 74 %
- Loss on drying (105°C):Max. 3 %
- Acidly reacting impurities:complying
- Ammonium (NH₄):Max. 0.01 %
- Silver (Ag):complying
- Arsenic (As):Max. 0.0002 %
- Copper (Cu):complying
- Lead:complying
- Lead (Pb):Max. 0.002 %
- Alkalis/earth alkalis:Max. 0.5 %
- Substances not precipitated by NH₄OH (SO₄):Max. 0.5 %
- Carbonate (CO₃):complying
- Chloride (Cl):Max. 0.02 %
- Sulfate (SO₄):Max. 0.03 %
- Residual solvents:complying