Ammonium heptamolybdate tetrahydrate 81.0-83.0% (MoO₃ basis) ACS, Fluka™
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:12054-85-2
:12027-67-7
Formula:(NH₄)₆Mo₇O₂₄·4H₂O
MDL Number:MFCD00167059
:1235.86 g/mol
Density:2.498 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:234-320-9
Specification Test Results
- Assay (as MoO₃):81.0 - 83.0 %
- Water insoluble matter:Max. 0.005 %
- Potassium (K):Max. 0.01 %
- Magnesium (Mg):Max. 0.005 %
- Sodium (Na):Max. 0.01 %
- Heavy metals (ICP) ACS:Max. 0.001 %
- Chloride (Cl):Max. 0.002 %
- Nitrate (NO₃):Max. 0.003 %
- Phosphate, Arsenate, Silicate (as SiO₂):Max. 0.001 %
- Phosphate (PO₄):Max. 5 ppm
- Sulfate (SO₄):Max. 0.02 %