
Ammonium cerium (IV) sulfate 0.1 M Ce(NH4)4(SO4)4 in aqueous solution Reag. Ph. Eur. volumetric solution, Fluka™
Catalog # BJ37941-1L
Supplier: Honeywell Research Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Size:1 L
- Cat. No.:BJ37941-1L
Specifications
Specification Test Results
- Factor:0.999 - 1.001
Specification Test Results
About this item
:
:596.52
Density:1.11 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:3264
ADR:8,III
:GHS05
CAS Number:7637-03-8
Formula:H₁₆CeN₄O₁₆S₄
MDL Number:MFCD00148851