

Ammonium cerium(IV) nitrate 0.1 M Ce(NH4)2(NO3)6 in aqueous solution ≥97% Reag. Ph. Eur. volumetric solution, Fluka™
BJ38187-1L
:
- Pack type:Plastic bottle
- Size:1 L
- Cat. No.:BJ38187-1L
- Supplier no.:38187-1L
- Appearance:complying
- Assay:0.095 - 0.105 mol/l
- Density (D 20/4):1.083 - 1.098
:
:548.22 g/mol
:1.1 g/cm³
:Ambient
:2796
:8,II
:GHS05
:GHS07
:16774-21-3
:(NH₄)₂Ce(NO₃)₆
:MFCD00151121