

Acetic acid 99.8-100.5%, puriss., meets analytical specification of BP, Ph. Eur., USP, FCC, Fluka™
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:64-19-7
:
Formula:H₃CCOOH
MDL Number:MFCD00036152
:60.05 g/mol
Boiling Pt:118 °C (1.013 hPa)
Melting Pt:16 °C
Density:1.05 g/cm³ (20 °C)
Flash Pt:40 °C
Storage Temperature:Ambient
EINECS:200-580-7
UN:2789
ADR:8,II
:GHS05
:GHS02
Specification Test Results
- Appearance:complying
- Assay:99.8 - 100.5 %
- Congealing - Freezing Point:min. 15.6 °C
- Non-volatile matter:max. 0.005 %
- Aluminium (Al):max. 0.00005 %
- Copper (Cu):max. 0.0005 %
- Iron (Fe):max. 0.0002 %
- Lead (Pb):max. 0.00005 %
- Zinc (Zn):max. 0.0005 %
- Heavy metals (as Pb):max. 0.0002 %
- Chloride (Cl):max. 0.0002 %
- Sulfate (SO₄):max. 0.0005 %
- KMnO₄ reducing matter (as HCOOH):max. 0.01 %
- KMnO₄ reducing substances:complying
- K₂Cr₂O₇ reducing matter:complying
- Residual solvents:complying
- Identity USP:complying