

Acetic acid 99-100%, Puriss., meets analytical specification of BP, Ph. Eur., USP
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:64-19-7
:
Formula:H₃CCOOH
MDL Number:MFCD00036152
:60.05 g/mol
Boiling Pt:118 °C (1.013 hPa)
Melting Pt:16 °C
Density:1.05 g/cm³ (20 °C)
Flash Pt:40 °C
Storage Temperature:Ambient
EINECS:200-580-7
UN:2789
ADR:8,II
:GHS05
:GHS02
Specification Test Results
- Assay:Min. 99 %
- Non-volatile matter:Max. 0.005 %
- Iron (Fe):Max. 0.0005 %
- Heavy metals (as Pb):Max. 0.0005 %
- Chloride (Cl):Max. 0.0005 %
- Sulfate (SO₄):Max. 0.0005 %
- Explosion Limit:16 %, 92 °F
- Vapor Density:2.07 (vs air)
- Autoignition Temperature:800 °F