Diesse Cube 30 Touch, Streck
Catalog # 76318-736
Supplier: Streck Laboratories
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Diesse CUBE 30 Touch Sed-Rate Analyzer
- Cat. No.:76318-736
Specifications
About this item
An automated instrument for high-volume erythrocyte sedmentation rate testing in EDTA tubes.
- 94% correlation to modified Westergren method
- Compatible with 13×75 mm (1.5 to 4.0 ml) K2EDTA or K3EDTA tubes
- Results in 20 minutes
- 30 Samples/Batch
- Internal barcode scanner and printer
- Automated patient and QC data archive
- Easily interfaces to LIS
- No reagents, no exposure to patient sample