EDTA disodium salt solution with zinc complex added (solution B) concentrate for dilution to 1L for hardness of water determinations, for complexometry (1 ml = 1°dH/100 ml H₂O), Fluka™
Catalog # BJ38056-1EA
Supplier: Honeywell Research Chemicals
CAS Number:
Specifications
- Pack type:Ampoule
- Size:1 Each
- Cat. No.:BJ38056-1EA
Specifications
Specification Test Results
- Factor:0.998 - 1.002
Specification Test Results
About this item
:
Boiling Pt:100 °C
Storage Temperature:Ambient
MDL Number:MFCD00147223