Disposable Macro Cuvette, FireflySci
3.50 ml, W×D 10×10 mm, W×D×H 12.5×12.5×45 mm, 10 mm, UV Quartz
Catalog # 76016-584
Supplier: FIREFLYSCI, INC.
Supplier #: 1FLPUV
Jump to:
Product Details & Documents
Cuvettes are for rapid assays; use one and throw it away.
Four windows and base optically polished.
Specifications
- Volume:3.50 ml
- Material:UV Quartz
- Lightpath:10 mm
- External dimensions:W×D×H 12.5×12.5×45 mm
- Internal dimensions:W×D 10×10 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



