Type 11FL Macro Fluorescence Cuvettes with Quartz Cap, Firefly Sci
Supplier: FIREFLYSCI, INC.
Certificates
About this item
This cell provides a transparent, airtight glass cap, which provides an airtight seal, ensured by the ground finish on the inside of the cap and the top of the cuvette.
- Fire fused to ensure the best bonding
- Lightpaths from 5 mm up to 40 mm available
- Four windows and the base optically polished
- Includes airtight glass cap
Hand made ensuring the highest quality optical polish with no defects. Every cell is thoroughly inspected before leaving the factory.