CE Multisurface Cleaner™, Cleaning Mop, Vileda Professional®
Catalog # 76318-342
Supplier: Vileda Professional - FHP
Specifications
- Description:CE Multisurface Cleaner™
- Material:Plastic/metal
- Size:23"
- Cat. No.:76318-342
Specifications
About this item
Designed to clean pipes, hoods, and other hard to reach areas or uneven surfaces with minimal effort and maximum efficiency. Versatile and easily bendable to give the tool different shapes to effectively clean any surface.
- Completely autoclavable and rust free
- Extremely versatile
- Easily bendable to a variety of different shapes to effectively clean any surface
- Sturdy enough to not bend in use
Customized tools for special surfaces. The easiest, most effective way to clean pipes, soft walls curtains, tanks and other hard to reach areas in Controlled Environments.
A new line of specialty above-the-floor tools by Vileda Professional®. This line is ideal for all surfaces that are hard or difficult to reach, clean, and disinfect. Specialty tools use Controlled Environment compatible disposable microfiber sleeves that are already validated materials and are an extension of a current CE floor cleaning system.
Certifications: Meet CE requirements up to ISO 4-9/GMP A/B, C, D.