ICP-MS Standard, Internal Standard, 6 analytes, Inorganic Ventures
Catalog # 103254-956
Supplier: Inorganic Ventures
CAS Number:
Specifications
- Matrix:3% HNO₃
- Description:ICP-MS Internal Standard
- Size:125 ml
- Cat. No.:103254-956
- Supplier no.:IVICPMS71D-125
Specifications
About this item
ICP-MS standard is a certified reference material set in a nitric acid matrix for stability.
6 analytes at 10 µg/ml: Bismuth, Scandium, Yttrium, Indium, Terbium, Lithium-6.
Certifications: Traceable to NIST. Manufactured in accordance with our stingent quality assurance guidelines. ISO 17025 and ISO 17034 certified.