Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
HPC Broth, GVS

HPC Broth, GVS

HPC Broth, GVS
Certificates

2 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

HPC broth and HPC broth with TTC Heterotrophic Plate Count (HPC) broths are used to determine live heterotrophs in drinking water and other media at incubation temperatures of 35 °C. All bacteria grow on HPC with indicator media and produce a red color. This is a result of the precipitation of formazan following the reduction of 2,3,5- TTC by bacteria.

Product Family Options

Product Information

  • Size
    Pack type
    Availability
    Price
  • 10057-678
    HPC Broth with TTC
    HPC Broth with TTC
    50×2 ml
    Ampoule
    Log in for availability
    Log in for availability
    Pack of 50 (2ml)
    Pack of 50 (2ml) - $216.53
    $216.53
    Pack of 50 (2ml)
       $216.53

    Supplier Number: 10496151
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • HPC Broth
    50×2 ml
    Ampoule
    Log in for availability
    Log in for availability
    Pack of 50 (2ml)
    Pack of 50 (2ml) - $176.41
    $176.41
    Pack of 50 (2ml)
       $176.41

    Supplier Number: 10496164
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.