Hibernate E Media, BrainBits®
Hibernate E
Catalog # MSPP-HE
Marketsource
Supplier: BRAINBITS LLC MS
Supplier #: HE-500
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Hibernate® is a nutrient medium for the maintenance of neural tissue or cells in ambient carbondioxide levels (US PATENT 6,180,404).
Hibernate-E is used for embryonic tissue.
Specifications
Specifications
- Description:Hibernate E