Orders LinkContactUsLinkComponent
Glass Evaporating Basins
Supplier:   Wards
Glass Evaporating Basins
Supplier:  Wards
Certificates
Glass Evaporating Basins
Glass Evaporating Basins
7 Options Available Below
Product Details & Documents
Ideal for drying applications in microwave and ovens.
Evaporating basins made of borosilicate glass. The basins have a flat bottom with spout for easy pouring.
Eisco Labs borosilicate glass has the following heat tolerances:
Strain Point: –515 °C
Annealing Point: 565 °C
Softening Point: 820 °C
Product Family Options
Product Information
  • Capacity
    Ø
    Height
    Availability
    Price
  • 15 ml
    0.9" (Base), 2" (Top)
    1.2"
    Log in for availability
    Log in for availability
    Each
    Each - $10.15
    $10.15
    Each
       $10.15
    Supplier Number: CH0068A01
  • 45 ml
    1.2" (Base), 2.4" (Top)
    1.9"
    Log in for availability
    Log in for availability
    Each
    Each - $12.50
    $12.50
    Each
       $12.50
    Supplier Number: CH0068A02
  • 100 ml
    1.4" (Base), 3.1" (Top)
    1"
    Log in for availability
    Log in for availability
    Each
    Each - $17.90
    $17.90
    Each
       $17.90
    Supplier Number: CH0068A
  • 250 ml
    3"
    1.3"
    Log in for availability
    Log in for availability
    Each
    Each - $36.29
    $36.29
    Each
       $36.29
    Supplier Number: CH0068B
  • 500 ml
    2.3" (Base), 5.2" (Top)
    2.5"
    Log in for availability
    Log in for availability
    Each
    Each - $32.55
    $32.55
    Each
       $32.55
    Supplier Number: CH0068B01
  • 600 ml
    2.3" (Base), 5.9" (Top)
    3.4"
    Log in for availability
    Log in for availability
    Each
    Each - $53.05
    $53.05
    Each
       $53.05
    Supplier Number: CH0068C
  • 2000 ml
    3.6" (Base), 8" (Top)
    4"
    Log in for availability
    Log in for availability
    Each
    Each - $80.15
    $80.15
    Each
       $80.15
    Supplier Number: CH0068D