Florisil SPE Columns, Honeywell Burdick & Jackson
Supplier: Honeywell Research Chemicals
Certificates
About this item
These SPE columns are made from medical-grade polypropylene with PE frits.
- Wide selection of support materials and multiple cartridge/column sizes
- Every stationary phase batch tested to ensure lot-to-lot reproducibility
- Reliable quality
All columns are normal phase and should be preconditioned with a non-polar solvent with the elution solvent being polar.
Specifications
- Catalog No:
- BJ9111
- BJ9100
- BJ9106
- BJ9104
- BJ9102
- Endcapped:
- N/A
- N/A
- N/A
- Storage:
- Ambient
- Ambient
- Ambient
- Ambient
- Ambient