Order Entry
Puerto Rico
ContactUsLinkComponent
 
 
Bromate-Bromide 0.25 M Br2 (0.5 N) volumetric solution, Fluka™
  BJ35007-1L
 :  
 
Bromate-Bromide 0.25 M Br2 (0.5 N) volumetric solution, Fluka™
  BJ35007-1L
 :  35007-1L
 :  

 

  • Pack type:
    Glass bottle
  • Size:
    1 L
  • Cat. No.:
    BJ35007-1L

 

 

  • Factor:
    0.998 - 1.002

 

 

 :
 :1.05 g/cm³
 :GHS08
 :Ambient
 :7758-02-3
 :KBrO₃/KBr