Sulfonazo III sodium salt indicator for titration (SO4), Fluka™
Catalog # BJ86170-5G
Supplier: Honeywell Research Chemicals
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Plastic bottle
- Size:5 g
- Cat. No.:BJ86170-5G
Specifications
Specification Test Results
- Appearance:complying
- Identity (IR):complying
- Solution 1g to 100ml (H₂O):complying
- Loss on drying (110°C):Max. 12 %
- Application test:complying
- Absorption maximum:625 - 635 nm
- Absorbtion (1%/1cm):Min. 80
Specification Test Results
About this item
:
:776.55 g/mol
Storage Temperature:Ambient
CAS Number:68504-35-8
Formula:C₂₂H₁₆N₄O₁₄S₄
MDL Number:MFCD00003942