Ethidium Homodimer III (EthD-III) 1 mM in DMSO used in nuclear staining
Catalog # 89139-130
Supplier: Biotium
CAS Number:
Specifications
- Size:200 µl
- Cat. No.:89139-130
- Supplier no.:40051
Specifications
About this item
:
:~1000 g/mol
Storage Temperature:Refrigerator